Learn More
AbboMax Morphine Monoclonal Antibody (ABM063)
Description
Positive control: HIV infected tissue Cellular location: Cell Membrane.
Specifications
Specifications
| Antigen | Morphine |
| Applications | ELISA, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | ABM063 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with proprietary stabilizer and 0.01% sodium azide |
| Gene Alias | C17H19NO3; Morphia; Morphin; Morphinum; Morphium |
| Host Species | Mouse |
| Immunogen | Morphine chemcially conjugated to KLH. |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.