Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Morphine Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501984439
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-4439 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-4439 Supplier Bioss Supplier No. BS2029R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Morphine Base is an opiate alkaloid isolated from the plant Papaver somniferum and produced synthetically. Morphine binds to and activates specific opiate receptors (delta, mu and kappa), each of which are involved in controlling different brain functions. In the central nervous and gastrointestinal systems, this agent exhibits widespread effects including analgesia, anxiolysis, euphoria, sedation, respiratory depression, and gastrointestinal system smooth muscle contraction. [National Center for Biotechnology Information. PubChem Compound Database; CID=5288826].

Specifications

Antigen Morphine
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Alias C17H19NO3; Morphia; Morphin; Morphinum; Morphium
Host Species Rabbit
Immunogen KLH conjugated Morphine.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.