Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Morpholine-4-carboxamidine hydrobromide, 98%

Catalog No. p-7024776
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 157415-17-3 | C5H12BrN3O | 210.075 g/mol

It is used as an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an active pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Hygroscopic. Store away from oxidizing agents, water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 157415-17-3
Molecular Formula C5H12BrN3O
Molecular Weight (g/mol) 210.075
MDL Number MFCD00051994
InChI Key IOHSDHMCZXCESZ-UHFFFAOYSA-N
Synonym morpholine-4-carboximidamide hydrobromide, morpholinoformamidine hydrobromide, 4-morpholinylformamidine hydrobromide, morpholine-4-carboxamidine hydrobromide, 4-morpholinecarboximidamide, monohydrobromide, acmc-20adpt, morpholine-4-carboxamidine, bromide, morpholin-4-carboxamidine hydrobromide
PubChem CID 2806564
IUPAC Name morpholine-4-carboximidamide;hydrobromide
SMILES C1COCCN1C(=N)N.Br

Specifications

Melting Point ∼155°C to 165°C
Quantity 5 g
Beilstein 7389038
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 210.08
Percent Purity 98%
Chemical Name or Material Morpholine-4-carboxamidine hydrobromide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.