Learn More
Morpholine, 99%
Description
Morpholine is a common additive for pH adjustment in both fossil fuel and nuclear power plant steam systems. It is widely used in organic synthesis as a building block in the preparation of the antibiotic linezolid, the anticancer agent gefitinib and the analgesic dextromoramide. It is also used as a solvent in chemical reactions and in the preparation of enamines. Further, it serves as a chemical emulsifier in the process of waxing fruit.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Morpholine is a common additive for pH adjustment in both fossil fuel and nuclear power plant steam systems. It is widely used in organic synthesis as a building block in the preparation of the antibiotic linezolid, the anticancer agent gefitinib and the analgesic dextromoramide. It is also used as a solvent in chemical reactions and in the preparation of enamines. Further, it serves as a chemical emulsifier in the process of waxing fruit.
Solubility
Miscible with water.
Notes
Hygroscopic. Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | °C to 5°C |
| Density | 1.007 g/mL |
| Boiling Point | 129°C |
| Flash Point | 35°C (95°F) |
| Assay Percent Range | 99% |
| Refractive Index | 1.454 |
| Quantity | 500 g |
| UN Number | UN2054 |
| Beilstein | 102549 |
| Sensitivity | Hygroscopic |
| Show More |
Safety and Handling
| Hazard Category | H226-H302-H311+H331-H314-H335-H373 |
| Hazard Statement | GHS H Statement H314-H226-H302-H312-H332 |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: I; Proper Shipping Name: MORPHOLINE |
| EINECSNumber | 203-815-1 |
| RTECSNumber | QD6475000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.