Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MOS Antibody (R38.1), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB1002739AF594
Description
MOS Monoclonal specifically detects MOS in Xenopus samples. It is validated for Western Blot, ELISA.Specifications
MOS | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
MOS | |
MBP-mos fusion protein. | |
0.1 mL | |
Primary | |
Xenopus species | |
IgG2b |
Western Blot, ELISA | |
R38.1 | |
Western Blot, ELISA | |
c-mos, EC 2.7.11, EC 2.7.11.1, MGC119962, MGC119963, MSV, oncogene MOS, Moloney murine sarcoma virus, Oocyte maturation factor mos, Proto-oncogene c-Mos, proto-oncogene serine/threonine-protein kinase mos, v-mos Moloney murine sarcoma viral oncogene homolog | |
Mouse | |
Protein A purified | |
Cell Cycle and Replication | |
4342.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction