Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MOS Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544590
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544590 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544590 Supplier Invitrogen™ Supplier No. PA544590
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: LNVARLRHDN IVRVVAASTR TPAGSNSLGT IIMEFGGNVT LHQVIYGAAG Sequence homology: Cow: 92%; Dog: 100%; Guinea Pig: 86%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 93%; Rat: 100%.

Protein kinases are enzymes that transfer a phosphate group from a phosphate donor, generally the g phosphate of ATP, onto an acceptor amino acid in a substrate protein. By this basic mechanism, protein kinases mediate most of the signal transduction in eukaryotic cells, regulating cellular metabolism, transcription, cell cycle progression, cytoskeletal rearrangement and cell movement, apoptosis, and differentiation. With more than 500 gene products, the protein kinase family is one of the largest families of proteins in eukaryotes. The family has been classified in 8 major groups based on sequence comparison of their tyrosine (PTK) or serine/threonine (STK) kinase catalytic domains. The STE group (homologs of yeast Sterile 7, 11, 20 kinases) consists of 50 kinases related to the mitogen-activated protein kinase (MAPK) cascade families (Ste7/MAP2K, Ste11/MAP3K, and Ste20/MAP4K). MAP kinase cascades, consisting of a MAPK and one or more upstream regulatory kinases (MAPKKs) have been best characterized in the yeast pheromone response pathway. Pheromones bind to Ste cell surface receptors and activate yeast MAPK pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MOS
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene MOS
Gene Accession No. P00540
Gene Alias C-MOS; c-mos oncogene; c-mos proto-oncogene; MGC119962; MGC119963; Moloney sarcoma oncogene; mos; MOS proto-oncogene, serine/threonine kinase; MOS proto-oncogene, serine/threonine kinase L homeolog; mos.L; mosxe; MSV; oncogene MOS, Moloney murine sarcoma virus; oocyte maturation factor mos; p39; p39-mos; pp39-mos; protein kinase; proto-oncogene c-Mos; Proto-oncogene serine/threonine-protein kinase mos; serine/threonine-protein kinase mos; v-mos Moloney murine sarcoma viral oncogene homolog; v-mos Moloney murine sarcoma viral oncogene homolog L homeolog; XELAEV_18032032mg
Gene Symbols MOS
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human MOS.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4342
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.