Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ MOSC1 Overexpression Lysate
SDP

Catalog No. NBL113189 Shop All Novus Biologicals Products
Catalog No. NBL113189 Supplier R&D Systems™ Supplier No. NBL113189

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems.

The MOSC1 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.

Specifications

Cell Line The lysate was created in HEK293T cells, using plasmid ID RC23674 and based on accession number NM_22746. The protein contains a C-terminal DDK tag.
Storage Buffer RIPA buffer
Purity Protein
Test Specificity Homo sapiens MOCO sulphurase C-terminal domain containing 1 (MOSC1), mRNA.
Content And Storage -80°C. Avoid freeze-thaw cycles.
Product Line Novus
Protein State Native
For Use With (Application) Western Blot

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.