Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MOSC1 Polyclonal Antibody

Catalog No. PIPA5121771
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121771 20 μL
PIPA5119290 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5121771 Supplier Invitrogen™ Supplier No. PA5121771
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MOSC1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

MOSC1 (MOCO sulphurase C-terminal domain containing 1), also known as MARC1, is a 337 amino acid mitochondrial protein that is thought to function as an oxidoreductase. Existing as three alternatively spliced isoforms, MOSC1 contains one MOSC domain and binds molybdenum as a cofactor. The gene encoding MOSC1 maps to human chromosome 1, which spans 260 million base pairs, contains over 3,000 genes and comprises nearly 8% of the human genome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MOSC1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene MARC1
Gene Accession No. Q5VT66, Q9CW42
Gene Alias mARC1; mitochondrial amidoxime reducing component 1; mitochondrial amidoxime-reducing component 1; moco sulfurase C-terminal domain-containing protein 1; MOCO sulphurase C-terminal domain containing 1; molybdenum cofactor sulfurase C-terminal domain-containing protein 1; MOSC domain-containing protein 1; MOSC domain-containing protein 1, mitochondrial; MOSC1
Gene Symbols MARC1
Host Species Rabbit
Immunogen Recombinant protein MOCO Sulphurase C-Terminal Domain Containing Protein 1. The antigen corresponds to amino acid range 41-335 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 537028, 64757, 66112, 690745
Target Species Bovine, Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.