Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MOSC2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 4 publications
$405.00 - $670.00
Specifications
Antigen | MOSC2 |
---|---|
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
MOSC2 Polyclonal specifically detects MOSC2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
MOSC2 | |
Polyclonal | |
Rabbit | |
Human | |
FLJ20605, MOCO sulphurase C-terminal domain containing 2, MOSC domain-containing protein 2, mitochondrial, RP11-270A6.1 | |
43161 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
54996 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:FQVAYPDYCPLLIMTDASLVDLNTRMEKKMKMENFRPNIVVTGCDAFEEDTWDELLIGSVEVKKVMACPRCILTTVDPDTGVIDRKQPLDTLKSYRL | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title