Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MOSC2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553276
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553276 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553276 Supplier Invitrogen™ Supplier No. PA553276
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FQVAYPDYCP LLIMTDASLV DLNTRMEKKM KMENFRPNIV VTGCDAFEED TWDELLIGSV EVKKVMACPR CILTTVDPDT GVIDRKQPLD TLKSYRL Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 74%.

As a component of the benzamidoxime prodrug-converting complex required to reduce N-hydroxylated prodrugs, such as benzamidoxime. Also able to reduce N(omega)-hydroxy-L-arginine (NOHA) and N(omega)-hydroxy-N(delta)-methyl-L-arginine (NHAM) into L-arginine and N(delta)-methyl-L-arginine, respectively.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MOSC2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MARC2
Gene Accession No. Q969Z3
Gene Alias 2810484M10Rik; mARC1; MARC2; Mg87; Mitochondrial amidoxime reducing component 2; moco sulfurase C-terminal domain-containing protein 2; MOCO sulphurase C-terminal domain containing 2; molybdenum cofactor sulfurase C-terminal domain-containing protein 2; MOSC domain-containing protein 2; MOSC domain-containing protein 2, mitochondrial; Mosc2; RP11-270A6.1
Gene Symbols MARC2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MOSC2. Recombinant protein control fragment (Product #RP-90153).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54996
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.