Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MOSPD1 Antibody, Alexa Fluor™ 532, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP281739AF532
This item is not returnable.
View return policy
Description
MOSPD1 Polyclonal antibody specifically detects MOSPD1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
MOSPD1 | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Peptide affinity purified | |
RUO | |
56180 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Alexa Fluor 532 | |
dJ473B4, motile sperm domain containing 1, motile sperm domain-containing protein 1 | |
MOSPD1 antibody was raised against a 19 amino acid peptide near the center of human MOSPD1 . The immunogen is located within amino acids 110 - 160 of MOSPD1. Amino Acid Squence: LPSAKEQQKEEEEKRLKEH | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction