Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MOSPD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556659
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556659 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556659 Supplier Invitrogen™ Supplier No. PA556659
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NLPVFVFPTE LIFYADDQST HKQVLTLYNP YEFALKFKVL CTTPNKYVVV DAAGAVKPQC CVDIVIRHRD VRSCHYGVID KFRLQVSEQS QRKALGRKEV VATLLPSAKE QQKEEEEKRL KEHLTESLFF EQSFQPENRA VSS Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.

The Motile sperm domain-containing protein 1 (MOSPD1) is part of a family of proteins defined by the presence of a major sperm protein (MSP) domain and two transmembrane domains. MOSPD1 codes for a small protein that localizes to the endoplasmic reticulum (ER) and the Golgi apparatus and has been suggested to play a role in the developmental regulation at the switch between mesenchymal and epithelial cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MOSPD1
Applications Western Blot, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MOSPD1
Gene Accession No. Q9UJG1
Gene Alias 1810018L05Rik; DJ473B4; Mospd1; motile sperm domain containing 1; motile sperm domain-containing protein 1
Gene Symbols MOSPD1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MOSPD1. Recombinant protein control fragment (Product #RP-92180).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56180
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.