Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MOSPD2 Mouse anti-Human, Clone: 1MOS‐4E10, MilliporeSigma™

Catalog No. MABF2202100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF2202100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF2202100 Supplier MilliporeSigma Supplier No. MABF2202100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MOSPD2 Monoclonal antibody specifically detects MOSPD2 in Human samples. It is validated for Immunohistochemistry, Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MOSPD2
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 1MOS‐4E10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Host Species Mouse
Immunogen A recombinant fragment corresponding to 119 amino acid MSP domain of human Motile sperm domain-containing protein 2 (MOSPD2).
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.