Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Motilin Antibody, Allophycocyanin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297102APC
Description
Motilin Polyclonal antibody specifically detects Motilin in Human samples. It is validated for ELISASpecifications
Motilin | |
Polyclonal | |
ELISA | |
MGC138519, motilin, prepromotilin, promotilin | |
Produced in rabbits immunized with purified, recombinant Human Motilin (Accession#: AAH69675.1; Met1-Lys115) | |
0.1 mL | |
Signal Transduction | |
4295 | |
Store at 4C in the dark. | |
IgG |
ELISA | |
APC | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction