Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Motilin Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA564415
Description
Immunogen sequence: AFVPIFTYGE LQRMQEKERN KGQKKSLSVW QRSGEEGPVD PAEPIREEEN Highest antigen sequence identity to the following orthologs: Mouse - 34%, Rat - 30%.
This gene encodes a small peptide hormone that is secreted by cells of the small intestine to regulate gastrointestinal contractions and motility. Proteolytic processing of the secreted protein produces the mature peptide and a byproduct referred to as motilin-associated peptide (MAP). Two transcript variants encoding different preproprotein isoforms but the same mature peptide have been found for this gene.
Specifications
Motilin | |
Polyclonal | |
Unconjugated | |
MLN | |
gastrin-related protein; MAP; MGC138519; MLN; Motilin; Motilin-associated peptide; prepromotilin; promotilin; secreted protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4295 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.20 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P12872 | |
MLN | |
Recombinant protein corresponding to Human Motilin. Recombinant protein control fragment (Product #RP-104166). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction