Description
- Designed for fast, accurate positioning during stereotaxic surgery
- Ideal for inexperienced users
- Motorized manipulator arms employ a touchscreen display for easy-to-control movement of each axis
- Manual movement available anytime
- Automatic motorized movement using touchscreen display
- Various speeds (3.0 mm/s to 0.15 μm/s*DV only)
- 10 μm movement
Specifications
Specifications
| For Use With (Application) | Stereotaxic Surgery |
| Animal | Rat |
| Includes | Base, u-frame, built-in warmer (controller extra), 3-axis motorized manipulator arm, touchscreen controller, corner-clamp probe holder, rat adaptor |