Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse 4-1BB Ligand/TNFSF9 Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF1246
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF1246 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF1246 Supplier R&D Systems Supplier No. BAF1246
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

4-1BB Ligand/TNFSF9 Polyclonal specifically detects 4-1BB Ligand/TNFSF9 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen 4-1BB Ligand/TNFSF9
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL
Gene Accession No. P41274
Gene Alias 4-1BB ligand, 41BB Ligand, 4-1BBL, 4-1BB-L, CD137L, homolog of mouse 4-1BB-L, receptor 4-1BB ligand, TNFSF9, tumor necrosis factor (ligand) superfamily, member 9, tumor necrosis factor ligand superfamily member 9
Gene Symbols TNFSF9
Host Species Goat
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse 4-1BB Ligand/TNFSF9 Arg104-Glu309 Accession # P41274
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8744
Test Specificity Detects mouse 4-1BB Ligand/TNFSF9 in Western blots. In Western blots, less than 5% cross-reactivity with recombinant mouse (rm) Fas Ligand, rmOX40 Ligand, rmTWEAK, rmTRAIL, and rmTNF-alpha is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.