Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse 4-1BB/TNFRSF9/CD137 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB937
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB937 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB937 Supplier R&D Systems Supplier No. MAB937
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

4-1BB/TNFRSF9/CD137 Monoclonal specifically detects 4-1BB/TNFRSF9/CD137 in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen 4-1BB/TNFRSF9/CD137
Applications Western Blot, Flow Cytometry, CyTOF, ELISA
Classification Monoclonal
Clone 158332
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P20334
Gene Alias 41BB, 4-1BB, 4-1BB ligand receptor, CD137, CD137 antigen, CD137MGC2172, CDw137, FLJ43501, ILA, ILAhomolog of mouse 4-1BB, induced by lymphocyte activation (ILA), interleukin-activated receptor, homolog of mouse Ly63, receptor protein 4-1BB, T cell antigen ILA, T-cell antigen 4-1BB homolog, T-cell antigen ILA, TNFRSF9, tumor necrosis factor receptor superfamily member 9, tumor necrosis factor receptor superfamily, member 9
Gene Symbols TNFRSF9
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse 4-1BB Val24-Leu187 Accession # P20334
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3604
Test Specificity Detects mouse 4-1BB in direct ELISAs and Western blots. Does not cross-react with recombinant human (rh) 4-1BB, rhCD27, recombinant mouse (rm) CD27, rhCD30, rmCD30, rhCD40, rmCD40, rhDR3, rhDR6, rhEDAR, rmEDAR, rhFas, rmFas, rhGITR, rmGITR, rhHVEM, rhLTR beta , rhNGF R, rhOPG, rmOPG, rhRANK, rmRANK, rhTROY, rmTROY, rhTNF sRI, rhTNF sRII, or rmTNF sRII.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.