Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse ACE/CD143 Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF1513
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF1513 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF1513 Supplier R&D Systems Supplier No. BAF1513
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

ACE/CD143 Polyclonal specifically detects ACE/CD143 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, ELISA Detection (Matched Antibody Pair).

Specifications

Antigen ACE/CD143
Applications Western Blot, Flow Cytometry, Immunohistochemistry, ELISA
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 5-15 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL
Gene Accession No. P09470
Gene Alias ACE1angiotensin converting enzyme, somatic isoform, angiotensin I converting enzyme (peptidyl-dipeptidase A) 1, carboxycathepsin, CD143, CD143 antigen, DCP, DCP1, DCP1angiotensin-converting enzyme, dipeptidyl carboxypeptidase 1, Dipeptidyl carboxypeptidase I, EC 3.2.1.-, EC 3.4.15.1, Kininase II, MGC26566, MVCD3, peptidase P, testicular ECA
Gene Symbols ACE
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse ACE/CD143 Leu35-Gln1264 Accession # P09470
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1636
Test Specificity Detects mouse ACE/CD143 in ELISAs and Western blots. In sandwich immunoassays, less than 0.2% cross-reactivity with recombinant mouse ACE-2 and recombinant human ACE is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.