Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse ADAM15 Ectodomain Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $643.50
Specifications
Antigen | ADAM15 |
---|---|
Clone | 140104 |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Classification | Monoclonal |
Description
ADAM15 Monoclonal specifically detects ADAM15 in Mouse samples. It is validated for Western Blot.Specifications
ADAM15 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
ADAM 15, ADAM metallopeptidase domain 15, disintegrin-like, and cysteine-rich protein 15, EC 3.4.24, MDC15, MDC-15, Metargidin | |
ADAM15 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
140104 | |
Western Blot | |
Unconjugated | |
Rat | |
Mouse | |
O88839 | |
8751 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse ADAM15 Arg18-Ser694 Accession # O88839 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse ADAM15 Ectodomain in Western blots. In Western blots, no cross-reactivity with recombinant human ADAM8, 9, 15, 17, or recombinant mouse ADAM10 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title