Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse ADAM19 Ectodomain Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB1229SP
Change view
Click to view available options
Quantity:
25 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1229SP 25 μg
MAB1229 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB1229SP Supplier R&D Systems Supplier No. MAB1229SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

ADAM19 Monoclonal specifically detects ADAM19 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen ADAM19
Applications Western Blot
Classification Monoclonal
Clone 164646
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. O35674
Gene Alias a disintegrin and metalloproteinase domain 19 (meltrin beta), ADAM 19, ADAM metallopeptidase domain 19, disintegrin and metalloproteinase domain-containing protein 19, EC 3.4.24, EC 3.4.24.-, MADDAM, Meltrin beta, Meltrin-beta, Metalloprotease and disintegrin dendritic antigen marker, MLTNBmeltrin beta
Gene Symbols ADAM19
Host Species Rat
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse ADAM19 Ectodomain Asp205-Gly705 (predicted) Accession # O35674
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8728
Test Specificity Detects mouse ADAM19 Ectodomain in direct ELISAs and Western blots. It also recognizes a C-terminal fragment (amino acids 588-705) of recombinant mouse (rm) ADAM19 in Western blots. In Western blots, no cross-reactivity with rmADAM10 or rmADAM15 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.