Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse ADAM33 Ectodomain Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $468.00
Specifications
Antigen | ADAM33 |
---|---|
Clone | 219515 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL |
Applications | Western Blot, Immunoprecipitation |
Description
ADAM33 Monoclonal specifically detects ADAM33 in Mouse samples. It is validated for Western Blot, Immunoprecipitation.Specifications
ADAM33 | |
LYOPH | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Rat | |
Mouse | |
Q923W9 | |
80332 | |
Mouse myeloma cell line NS0-derived recombinant mouse ADAM33 Ectodomain Glu205-Ala702 Accession # Q923W9 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse ADAM33 Ectodomain in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant mouse ADAM9, 10, 15, recombinant human (rh) ADAM8, rhTACE/ADAM17, rhADAM19, or rhADAM33 is observed. |
219515 | |
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
a disintegrin and metalloprotease 33, a disintegrin and metalloproteinase domain 33, ADAM 33, ADAM metallopeptidase domain 33, C20orf153, chromosome 20 open reading frame 153, disintegrin and metalloproteinase domain-containing protein 33, disintegrin and reprolysin metalloproteinase family protein, DJ964F7.1, DKFZp434K0521, EC 3.4.24.-, FLJ35308, FLJ36751, MGC149823, MGC71889 | |
ADAM33 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title