Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Adiponectin/Acrp30 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 3 publications
$200.50 - $2020.00
Specifications
Antigen | Adiponectin/Acrp30 |
---|---|
Clone | 173205 |
Concentration | LYOPH |
Dilution | Western Blot 2 ug/mL |
Applications | Western Blot |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB1119SP
![]() |
R&D Systems
MAB1119SP |
25 μg |
Each for $200.50
|
|
|||||
MAB1119100
![]() |
R&D Systems
MAB1119100 |
100 μg |
Each for $549.50
|
|
|||||
MAB1119500
![]() |
R&D Systems
MAB1119500 |
500 μg |
Each for $2,020.00
|
|
|||||
Description
Adiponectin/Acrp30 Monoclonal specifically detects Adiponectin/Acrp30 in Mouse samples. It is validated for Western Blot.Specifications
Adiponectin/Acrp30 | |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
ACDC, Acrp30, ACRP30ADPN, adipocyte, C1Q and collagen domain containing, adiponectin, adiponectin, C1Q and collagen domain containing, AdipoQ, ADIPQTL1, apM-1, ApM1, APM1APM-1, C1q and collagen domain-containing protein, GBP28, GBP28apM1, Gelatin-binding protein | |
ADIPOQ | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
173205 | |
Western Blot 2 ug/mL | |
Monoclonal | |
Rat | |
Mouse | |
Q60994 | |
9370 | |
Mouse myeloma cell line NS0-derived recombinant mouse Adiponectin Glu18-Asn247 Accession # Q60994 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects full-length mouse Adiponectin (aa 18-247) in direct ELISAs and Western blots. Shows no cross-reactivity with recombinant human Adiponectin. Does not detect the C-terminal globular domain of mouse Adiponectin (aa 111-247). |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title