Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Adiponectin/Acrp30 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 18 publications
Supplier: R&D Systems AF1119
This item is not returnable.
View return policy
Description
Adiponectin/Acrp30 Polyclonal specifically detects Adiponectin/Acrp30 in Mouse samples. It is validated for Western Blot, Immunocytochemistry.Specifications
Adiponectin/Acrp30 | |
Polyclonal | |
Unconjugated | |
Q60994 | |
ADIPOQ | |
Mouse myeloma cell line NS0-derived recombinant mouse Adiponectin/Acrp30 (R&D Systems, Catalog # 5095-AC) Glu18-Asn247 Accession # Q60994 | |
100 μg | |
Primary | |
Detects mouse Adiponectin/Acrp30 in direct ELISAs and Western blots. In direct ELISAs, approximately 50% cross-reactivity with recombinant rat Adiponectin is observed and approximately 30% cross-reactivity with recombinant human Adiponectin is observed. | |
Mouse | |
IgG |
Western Blot, Immunocytochemistry | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
ACDC, Acrp30, ACRP30ADPN, adipocyte, C1Q and collagen domain containing, adiponectin, adiponectin, C1Q and collagen domain containing, AdipoQ, ADIPQTL1, apM-1, ApM1, APM1APM-1, C1q and collagen domain-containing protein, GBP28, GBP28apM1, Gelatin-binding protein | |
Goat | |
Affinity Purified | |
RUO | |
9370 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction