Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Mouse Adiponectin/Acrp30 Quantikine ELISA Kit

Catalog No. p-4753770
Click to view available options
Quantity:
1 Plate
6 Plates
This item is not returnable. View return policy
This item is not returnable. View return policy

Mouse Adiponectin/Acrp30 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Mouse Adiponectin/Acrp30 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies mouse Adiponectin/Acrp30 in cell culture supernates (50 ul), tissue homogenates (50 ul), serum (10 ul), edta plasma (10 ul), heparin plasma (10 ul).

Specifications

Assay Range 0.16 - 10ng/mL
Assay Sensitivity 0.007ng/mL
Conjugate HRP
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type Solid Phase Sandwich ELISA
Sample Type Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum, Tissue Homogenates
Specificity Natural and Recombinant full-length Mouse Adiponectin. This assay only recognizes full-length Mouse Adiponectin. It does not recognize Mouse gAdiponectin.
Sufficient For 1 x 96-well Plate
Target Species Mouse
For Use With (Application) ELISA
Gene ID (Entrez) 11450
Quantity 1 Plate
Sample Volume Cell Culture Supernates (50uL),Tissue Homogenates (50uL),Serum (10uL),EDTA Plasma (10uL),Heparin Plasma (10uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym ACDC, Acrp30, ACRP30ADPN, adipocyte, C1Q and collagen domain containing, adiponectin, adiponectin, C1Q and collagen domain containing, AdipoQ, ADIPQTL1, apM-1, ApM1, APM1APM-1, C1q and collagen domain-containing protein, GBP28, GBP28apM1, Gelatin-binding protein
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.