Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse AHR Alexa Fluor 488-conjugated Antibody, R&D Systems™

Catalog No. IC6697G
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC6697G 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC6697G Supplier R&D Systems Supplier No. IC6697G
This item is not returnable. View return policy

Sheep Polyclonal Antibody

AHR Polyclonal specifically detects AHR in Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen AHR
Applications Flow Cytometry
Classification Polyclonal
Conjugate Alexa Fluor 488
Dilution Intracellular Staining by Flow Cytometry 5 uL/10^6 cells
Gene Accession No. P30561
Gene Alias Ah receptor, AhR, AH-receptor, aryl hydrocarbon receptor, BHLHE76, bHLHe76aromatic hydrocarbon receptor, Class E basic helix-loop-helix protein 76
Gene Symbols AHR
Host Species Sheep
Immunogen E. coli-derived recombinant mouse AHR Asn706-Ser805 (Thr758Ala) Accession # P30561
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 196
Test Specificity Detects human and mouse AHR in Western blots.
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.