Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse AHR Antibody, R&D Systems™

Sheep Polyclonal Antibody has been used in 2 publications
Supplier: R&D Systems AF6697
This item is not returnable.
View return policy
Description
AHR Polyclonal specifically detects AHR in Mouse samples. It is validated for Western Blot, Immunocytochemistry.Specifications
| AHR | |
| Polyclonal | |
| Unconjugated | |
| P30561 | |
| AHR | |
| E. coli-derived recombinant mouse AHR Asn706-Ser805 (Thr758Ala) Accession # P30561 | |
| 100 μg | |
| Primary | |
| Detects mouse AHR in Western blots. | |
| Mouse | |
| IgG |
| Western Blot, Immunocytochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| Ah receptor, AhR, AH-receptor, aryl hydrocarbon receptor, BHLHE76, bHLHe76aromatic hydrocarbon receptor, Class E basic helix-loop-helix protein 76 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 196 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction