Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse ALCAM/CD166 Alexa Fluor™ 750-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems FAB1172S100UG
This item is not returnable.
View return policy
Description
ALCAM/CD166 Monoclonal specifically detects ALCAM/CD166 in Mouse samples. It is validated for Flow Cytometry.Specifications
ALCAM/CD166 | |
Monoclonal | |
Alexa Fluor 750 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
activated leucocyte cell adhesion molecule, activated leukocyte cell adhesion moleculeFLJ38514, CD166, CD166 antigen, MEMDMGC71733 | |
Mouse myeloma cell line NS0-derived recombinant mouse ALCAM/CD166, Trp28-Lys527, Accession # AAC06342 | |
100 μg | |
Primary | |
Detects mouse ALCAM/CD166 in Western blots and direct ELISAs. In Western blots, shows 100% cross-reactivity with recombinant human (rh) ALCAM and no cross-reactivity with rhBCAM, rhEpCAM, recombinant mouse (rm) MAdCAM-1, rhMCAM, rmNCAM-L1, rmOCAM, or rmTROP-2. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG2a |
Flow Cytometry | |
200622 | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
AAC06342 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
214 | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction