Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse ALCAM/CD166 APC-conjugated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems FAB1172A
This item is not returnable.
View return policy
Description
ALCAM/CD166 Polyclonal specifically detects ALCAM/CD166 in Mouse samples. It is validated for Flow Cytometry.Specifications
ALCAM/CD166 | |
Polyclonal | |
Flow Cytometry 10 uL/10^6 cells | |
activated leucocyte cell adhesion molecule, activated leukocyte cell adhesion moleculeFLJ38514, CD166, CD166 antigen, MEMDMGC71733 | |
Goat | |
Affinity Purified | |
RUO | |
214 | |
Mouse | |
IgG |
Flow Cytometry | |
APC | |
AAC06342 | |
ALCAM | |
Mouse myeloma cell line NS0-derived recombinant mouse ALCAM/CD166 Trp28-Lys527 Accession # AAC06342 | |
100 Tests | |
Primary | |
Detects mouse ALCAM in direct ELISAs and Western blots. In direct ELISAs, approximately 40% cross-reactivity with recombinant human (rh) ALCAM is observed, and less than 10% cross-reactivity with rhBCAM, recombinant mouse (rm) OCAM, and rmMAdCAM-1 is observed. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction