Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse ALCAM/CD166 Fluorescein-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB1172F
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB1172F 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB1172F Supplier R&D Systems Supplier No. FAB1172F
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

ALCAM/CD166 Polyclonal specifically detects ALCAM/CD166 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen ALCAM/CD166
Applications Flow Cytometry
Classification Polyclonal
Conjugate Fluorescein
Dilution Flow Cytometry 10 uL/10^6 cells
Gene Accession No. AAC06342
Gene Alias activated leucocyte cell adhesion molecule, activated leukocyte cell adhesion moleculeFLJ38514, CD166, CD166 antigen, MEMDMGC71733
Gene Symbols ALCAM
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse ALCAM/CD166 Trp28-Lys527 Accession # AAC06342
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 214
Test Specificity Detects mouse ALCAM in direct ELISAs and Western blots. In direct ELISAs, approximately 40% cross-reactivity with recombinant human (rh) ALCAM is observed, and less than 10% cross-reactivity with rhBCAM, recombinant mouse (rm) OCAM, and rmMAdCAM-1 is observed.
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.