Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse AMIGO2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF2374
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF2374 100 μg
AF2374SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF2374 Supplier R&D Systems Supplier No. AF2374
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

AMIGO2 Polyclonal specifically detects AMIGO2 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen AMIGO2
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL
Gene Accession No. Q80ZD9
Gene Alias adhesion molecule with Ig-like domain 2, ALI1amphoterin induced gene 2, alivin 1, Alivin-1, AMIGO-2, DEGAamphoterin-induced protein 2, differentially expressed in gastric adenocarcinoma, Differentially expressed in gastric adenocarcinomas, transmembrane protein AMIGO2
Gene Symbols AMIGO2
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse AMIGO2 Gy38-Ala391 Accession # Q80ZD9
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 347902
Test Specificity Detects mouse AMIGO2 in direct ELISAs and Western blots. In direct ELISAs, approximately 40% cross-reactivity with recombinant human (rh) AMIGO2 is observed and less than 1% cross-reactivity with rhAMIGO1 and recombinant mouse AMIGO3 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.