Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse AMIGO3 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF2375SP
This item is not returnable.
View return policy
Description
AMIGO3 Polyclonal specifically detects AMIGO3 in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
AMIGO3 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
adhesion molecule with Ig-like domain 3, ALI3, Alivin-3, AMIGO-3, amphoterin-induced gene and ORF 3, amphoterin-induced protein 3, KIAA1851, MGC120552 | |
Goat | |
Affinity Purified | |
RUO | |
386724 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Flow Cytometry, CyTOF | |
LYOPH | |
Western Blot 0.1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
Q8C2S7 | |
AMIGO3 | |
Mouse myeloma cell line NS0-derived recombinant mouse AMIGO3 Thr20-Pro378 Accession # Q8C2S7 | |
25 μg | |
Primary | |
Detects mouse AMIGO3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 5% cross-reactivity with recombinant mouse AMIGO2 is observed and less than 2% cross-reactivity with recombinant human AMIGO1 is observed. | |
Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction