Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse AMIGO3 Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB2375
This item is not returnable.
View return policy
Description
AMIGO3 Monoclonal specifically detects AMIGO3 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
AMIGO3 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL | |
Q8C2S7 | |
AMIGO3 | |
Mouse myeloma cell line NS0-derived recombinant mouse AMIGO3 Thr20-Pro378 Accession # Q8C2S7 | |
100 μg | |
Primary | |
Detects mouse AMIGO3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse AMIGO2, recombinant human (rh) AMIGO1, or rhAMIGO2 is observed. | |
Mouse | |
Purified |
Western Blot, Immunohistochemistry | |
371301 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
adhesion molecule with Ig-like domain 3, ALI3, Alivin-3, AMIGO-3, amphoterin-induced gene and ORF 3, amphoterin-induced protein 3, KIAA1851, MGC120552 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
386724 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction