Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Aminopeptidase N/CD13 Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $468.00
Specifications
Antigen | Aminopeptidase N/CD13 |
---|---|
Clone | 945033 |
Dilution | Western Blot 1 ug/mL, Simple Western 10 ug/mL |
Applications | Western Blot |
Classification | Monoclonal |
Description
Aminopeptidase N/CD13 Monoclonal specifically detects Aminopeptidase N/CD13 in Mouse samples. It is validated for Western Blot, Simple Western.Specifications
Aminopeptidase N/CD13 | |
Western Blot 1 ug/mL, Simple Western 10 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
alanyl (membrane) aminopeptidase, Alanyl aminopeptidase, Aminopeptidase M, aminopeptidase N, ANPEP, AP-M, APN, AP-N, CD13, CD13 antigen, CD13APN, EC 3.4.11, EC 3.4.11.2, gp150, LAP1, Microsomal aminopeptidase, Myeloid plasma membrane glycoprotein CD13, P150, PEPN, PEPNhAPN | |
Anpep | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
945033 | |
Western Blot | |
Unconjugated | |
Rat | |
Mouse | |
P97449 | |
290 | |
NS0 mouse myeloma cell line transfected with mouse Aminopeptidase Lys69-Ser966 Accession # P97449 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse Aminopeptidase in direct ELISAs and Western blots. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title