Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Aminopeptidase PILS/ARTS1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB2500
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2500 100 μg
MAB2500SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB2500 Supplier R&D Systems Supplier No. MAB2500
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Aminopeptidase PILS/ARTS1 Monoclonal specifically detects Aminopeptidase PILS/ARTS1 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen Aminopeptidase PILS/ARTS1
Applications Western Blot
Classification Monoclonal
Clone 325908
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. NP_109636
Gene Alias Adipocyte-derived leucine aminopeptidase, A-LAP, A-LAPALAP, Aminopeptidase PILS, ARTS1, ARTS1aminopeptidase regulator of TNFR1 shedding, ARTS-1PILSAP, EC 3.4.11, EC 3.4.11.-, EC 3.4.11.1, endoplasmic reticulum aminopeptidase 1, endoplasmic reticulum aminopeptidase associated with antigen processing, ERAAP1, ERAP1, KIAA0525APPILS, PILS-AP, PILS-APERAAP, Puromycin-insensitive leucyl-specific aminopeptidase, Type 1 tumor necrosis factor receptor shedding aminopeptidase regulator
Gene Symbols ERAP1
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Aminopeptidase PILS/ARTS1 Ser27-Leu930 Accession # NP_109636
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51752
Test Specificity Detects mouse Aminopeptidase PILS/ARTS1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 10% cross-reactivity with recombinant human Aminopeptidase PILS/ARTS is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.