Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Angiopoietin-2 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF7186
This item is not returnable.
View return policy
Description
Angiopoietin-2 Polyclonal specifically detects Angiopoietin-2 in Mouse samples. It is validated for Western Blot.Specifications
Angiopoietin-2 | |
Polyclonal | |
Unconjugated | |
O35608 | |
ANGPT2 | |
Chinese hamster ovary cell line CHO-derived recombinant mouse Angiopoietin-2 Asp68-Phe496 Accession # O35608 | |
100 μg | |
Primary | |
Detects mouse Angiopoietin-2 in direct ELISAs and Western blots. In direct ELISAs, less than 2% cross-reactivity with recombinant human Angiopoietin-2, recombinant mouse (rm) Angiopoietin-1, and rmAngiopoietin-3 is observed. | |
Mouse | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.2 ug/mL | |
AGPT2, ANG2, ANG-2, angiopoietin 2, angiopoietin-2, angiopoietin-2a, angiopoietin-2B, angiopoitin 2, ANGPT2, Tie2-ligand | |
Sheep | |
Affinity Purified | |
RUO | |
285 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction