Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Angiopoietin-3 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF738
This item is not returnable.
View return policy
Description
Angiopoietin-3 Polyclonal specifically detects Angiopoietin-3 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Angiopoietin-3 | |
Polyclonal | |
Unconjugated | |
ANG3, ANG4, ANGPT3 | |
Goat | |
Affinity Purified | |
RUO | |
51378 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
Angpt4 | |
E. coli-derived recombinant mouse Angiopoietin-3 | |
100 μg | |
Primary | |
Detects mouse Angiopoietin-3 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) Angiopoietin-2 or rhAngoipoietin-3 was observed. | |
Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction