Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Angiopoietin-like 2 Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $655.00
Specifications
Antigen | Angiopoietin-like Protein 2/ANGPTL2 |
---|---|
Clone | 200001 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Description
Angiopoietin-like Protein 2/ANGPTL2 Monoclonal specifically detects Angiopoietin-like Protein 2/ANGPTL2 in Mouse samples. It is validated for Western Blot.Specifications
Angiopoietin-like Protein 2/ANGPTL2 | |
LYOPH | |
Western Blot | |
Unconjugated | |
Rat | |
Mouse | |
Q9R045 | |
23452 | |
E. coli-derived recombinant mouse Angiopoietin-like 2 Pro23-His493 Accession # Q9R045 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse Angiopoietin-like 2 in direct ELISAs and Western blots. In direct ELISAs, this antibody shows 100% cross-reactivity with recombinant human (rh) Angiopoietin-like 2 but no cross-reactivity with rhAngiopoietin-1, -2, -4, rmAngiopoietin-3, rhAngiopoietin-like 1, or rhAngiopoietin-like 7. |
200001 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
angiopoietin-like 2, Angiopoietin-like protein 2, angiopoietin-related protein 2, ANGPTL2, ANGRP2, ARP2HARP, MGC8889 | |
ANGPTL2 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title