Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Angiopoietin-like Protein 4/ANGPTL4 Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $467.50
Specifications
Antigen | Angiopoietin-like Protein 4/ANGPTL4 |
---|---|
Clone | 547113 |
Dilution | ELISA |
Applications | ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB97971SP
![]() |
R&D Systems
MAB97971SP |
25 μg |
Each for $200.50
|
|
|||||
MAB97971100
![]() |
R&D Systems
MAB97971100 |
100 μg |
Each for $467.50
|
|
|||||
Description
Angiopoietin-like Protein 4/ANGPTL4 Monoclonal antibody specifically detects Angiopoietin-like Protein 4/ANGPTL4 in Mouse samples. It is validated for ELISASpecifications
Angiopoietin-like Protein 4/ANGPTL4 | |
ELISA | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
Angiopoietin like Protein 4, angiopoietin-like 4, Angiopoietin-like protein 4, ANGPTL4, ARP4fasting-induced adipose factor, FIAF, FIAFhepatic angiopoietin-related protein, Hepatic fibrinogen/angiopoietin-related protein, HFARP, HFARPANGPTL2, NL2, peroxisome proliferator-activated receptor (PPAR) gamma inducedangiopoietin-related protein, PGAR, PGARangiopoietin-related protein 4, pp1158, PPARG angiopoietin related protein | |
CHO-derived mouse Angiopoietin-like Protein 4/ANGPTL4 protein,, Gln24-Ser410, Accession # Q9Z1P8 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
547113 | |
ELISA | |
Unconjugated | |
Rat | |
Mouse | |
Q9Z1P8 | |
51129 | |
IgG2a | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title