Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Angiopoietin-like Protein 4/ANGPTL4 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB9797100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB9797100 100 μg
MAB9797SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB9797100 Supplier R&D Systems Supplier No. MAB9797100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Angiopoietin-like Protein 4/ANGPTL4 Monoclonal antibody specifically detects Angiopoietin-like Protein 4/ANGPTL4 in Mouse samples. It is validated for ELISA

Specifications

Antigen Angiopoietin-like Protein 4/ANGPTL4
Applications ELISA
Classification Monoclonal
Clone 547112
Conjugate Unconjugated
Dilution ELISA
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS.
Gene Accession No. Q9Z1P8
Gene Alias Angiopoietin like Protein 4, angiopoietin-like 4, Angiopoietin-like protein 4, ANGPTL4, ARP4fasting-induced adipose factor, FIAF, FIAFhepatic angiopoietin-related protein, Hepatic fibrinogen/angiopoietin-related protein, HFARP, HFARPANGPTL2, NL2, peroxisome proliferator-activated receptor (PPAR) gamma inducedangiopoietin-related protein, PGAR, PGARangiopoietin-related protein 4, pp1158, PPARG angiopoietin related protein
Host Species Rat
Immunogen CHO-derived mouse Angiopoietin-like Protein 4/ANGPTL4 protein, Gln24-Ser410, Accession # Q9Z1P8
Purification Method Protein A or G purified from cell culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51129
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.