Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Angiopoietin-like Protein 7/ANGPTL7 Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB4960SP
This item is not returnable.
View return policy
Description
Angiopoietin-like Protein 7/ANGPTL7 Monoclonal specifically detects Angiopoietin-like Protein 7/ANGPTL7 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Angiopoietin-like Protein 7/ANGPTL7 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL | |
Q8R1Q3 | |
ANGPTL7 | |
Chinese hamster ovary cell line CHO-derived recombinant mouse Angiopoietin-like Protein 7/ANGPTL7 Gln22-Pro337 (predicted) Accession # Q8R1Q3 | |
25 μg | |
Primary | |
Detects mouse Angiopoietin-like Protein 7/ANGPTL7 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) Angiopoietin-like 1, 5, 7, recombinant mouse (rm) Angiopoietin-like 2, 3, 4, 6, rhAngiopoietin-1, rhAngiopoietin-4, rmAngiopoietin-2, or rmAngiopoietin-3 is observed. | |
Mouse | |
Purified |
Western Blot, Immunohistochemistry | |
538401 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
angiopoietin-like 7, Angiopoietin-like factor, Angiopoietin-like protein 7, angiopoietin-related protein 7, ANGPTL7, AngX, CDT6, CDT6angiopoietin-like factor (CDT6), Cornea-derived transcript 6 protein, dJ647M16.1, RP4-647M16.2 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
10218 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction