Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Angiopoietin-like Protein 7/ANGPTL7 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF4960
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF4960 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF4960 Supplier R&D Systems Supplier No. AF4960
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Angiopoietin-like Protein 7/ANGPTL7 Polyclonal specifically detects Angiopoietin-like Protein 7/ANGPTL7 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen Angiopoietin-like Protein 7/ANGPTL7
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Gene Accession No. Q8R1Q3
Gene Alias angiopoietin-like 7, Angiopoietin-like factor, Angiopoietin-like protein 7, angiopoietin-related protein 7, ANGPTL7, AngX, CDT6, CDT6angiopoietin-like factor (CDT6), Cornea-derived transcript 6 protein, dJ647M16.1, RP4-647M16.2
Gene Symbols ANGPTL7
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Angiopoietin-like Protein 7/ANGPTL7 Gln22-Pro337 Accession # Q8R1Q3
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10218
Test Specificity Detects mouse Angiopoietin-like Protein 7/ANGPTL7 in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant human ANGPTL7 is observed.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.