Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

17 beta-estradiol, Mouse, Clone: ESTR-1, Abnova™

Catalog No. 89135232 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-232 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-232 Supplier Abnova Corporation Supplier No. MAB2789
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against 17 beta-estradiol.

Specifications

Antigen 17 beta-estradiol
Applications ELISA
Classification Monoclonal
Clone ESTR-1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against 17 beta-estradiol.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Host Species Mouse
Immunogen 17 beta-Estradiol conjugated with BSA.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody recognizes 17 beta-estradiol-BSA conjugate and free estradiol. There is no cross-reactivity with BSA.
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.