Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3-OHKYN, Mouse, Clone: P3UI, Abnova™
Mouse monoclonal antibody raised against 3-OHKYN.
Supplier: Abnova Corporation MAB2404
Specifications
| 3-OHKYN | |
| Monoclonal | |
| Unconjugated | |
| Western Blot (2 ug/mL) The optimal working dilution should be determined by the end user. | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunocytochemistry, Western Blot | |
| P3UI | |
| Mouse monoclonal antibody raised against 3-OHKYN. | |
| In Tris-glycine, 150mM NaCl | |
| 3-OHKYN modified KLH. | |
| RUO | |
| This antibody is specific to 3-hydroxykynureinine modifications on N-alpha-acetyl lysine, N-alpha-acetyl histidine, N-alpha-acetyl arginine, and N-alpha-acetyl cysteine. It does not recognize the transaminase enzyme involved in the degradation of tryptophan. | |
| Store at -20°C or -80°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction