Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-HNE, Mouse, Clone: HNEJ-2, Abnova™

Catalog No. 89136100 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-100 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-100 Supplier Abnova Corporation Supplier No. MAB6115
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against 4-HNE.

Specifications

Antigen 4-HNE
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone HNEJ-2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against 4-HNE.
Dilution Immunohistochemistry (25 ug/mL) Western Blot (15 ug/mL) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from 50mM TBS (1% BSA)
Host Species Mouse
Immunogen 4-HNE-modified conjugated with KLH.
Quantity 20 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody shows almost negligible reactivity With proteins that were treated with other aldehydes such as: 2-nonenal, 2-hexenal, 1-hexenal, 4-hydroxy-2-hexenal, formaldehyde or glutaraldehyde.
Content And Storage Store at -20°C. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.