Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse anti-5'-Nucleotidase/CD73, Clone: 4G4, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 8 publications
Supplier: Novus Biologicals NBP1601350.05MG
Description
5'-Nucleotidase/CD73 Monoclonal specifically detects 5'-Nucleotidase/CD73 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen, Functional.Specifications
5'-Nucleotidase/CD73 | |
Monoclonal | |
0.1 mg/mL | |
Western Blot 1:100-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:50, Functional | |
P21589 | |
NT5E | |
Inflamed synovial stroma from rheumatoid arthitis patients | |
0.05 mg | |
Primary | |
Human | |
IgG1 |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
4G4 | |
Unconjugated | |
0.2 um filtered solution in PBS and 0.1% BSA with No Preservative | |
5' nucleotidase (CD73), 5'-NT, 5'-nucleotidase, ecto (CD73), CD73, CD73 antigen, E5NT, EC 3.1.3.5, ecto-5'-nucleotidase, eN, eNT, NT, NT55'-nucleotidase, NTE, Purine 5-Prime-Nucleotidase | |
Mouse | |
Affinity Purified | |
RUO | |
4907 | |
Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction