Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

8-hydroxy-guanosine, Mouse, Clone: 15A3, Abnova™

Catalog No. 89135078 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-078 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-078 Supplier Abnova Corporation Supplier No. MAB1998

Mouse monoclonal antibody raised against 8-hydroxy-guanosine.

Specifications

Antigen 8-hydroxy-guanosine
Applications ELISA, Immunohistochemistry
Classification Monoclonal
Clone 15A3
Conjugate Unconjugated
Formulation In 10mM Tris-Cl, 150mM NaCl, 1mM EDTA, pH 7.4 (0.09% sodium azide)
Host Species Mouse
Immunogen 8-hydroxy-guanosine conjugated with BSA and casein.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.