Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AATK, Mouse, Clone: 5B8, Abnova™

Catalog No. 89106123 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-106-123 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-106-123 Supplier Abnova Corporation Supplier No. H00009625M03A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant AATK.

The protein encoded by this gene contains a tyrosine kinase domain at the N-terminus and a proline-rich domain at the C-terminus. This gene is induced during apoptosis, and expression of this gene may be a necessary pre-requisite for the induction of growth arrest and/or apoptosis of myeloid precursor cells. This gene has been shown to produce neuronal differentiation in a neuroblastoma cell line. [provided by RefSeq

Sequence: SPEFVLKEAQEGCEPQAFAELASEGEGPGPETRLSTSLSGLNEKNPYRDSAYFSDLEAEAEATSGPEKKCGGDRAPGPELGLRSTGQPSEQVCLRPGVSG

Specifications

Antigen AATK
Applications ELISA, Western Blot
Classification Monoclonal
Clone 5B8
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant AATK.
Formulation ascites with no preservative
Gene AATK
Gene Accession No. BC047378
Gene Alias AATYK/AATYK1/KIAA0641/LMR1/LMTK1/p35BP
Gene Symbols AATK
Host Species Mouse
Immunogen AATK (AAH47378,161 a.a. ∽ 260 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9625
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.