Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abca1, Mouse, Clone: HJ1, Abnova™

Catalog No. 89110933 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-933 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-933 Supplier Abnova Corporation Supplier No. MAB2326
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant Abca1.

Specifications

Antigen Abca1
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone HJ1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant Abca1.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:400) Western Blot (1:2000) The optimal working dilution should be determined by the end user.
Formulation In Tris-glycine, 150mM NaCl (0.05% sodium azide)
Gene Abca1
Gene Alias Abc1
Gene Symbols Abca1
Host Species Mouse
Immunogen Recombinant protein corresponding to N-terminus extracellular region of mouse Abca1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11303
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C for short term. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.