Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ABCC1, Mouse, Clone: QCRL-3, Abnova™

Catalog No. 89112150 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-150 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-150 Supplier Abnova Corporation Supplier No. MAB5157
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant ABCC1.

The protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra-and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This full transporter is a member of the MRP subfamily which is involved in multi-drug resistance. This protein functions as a multispecific organic anion transporter, with oxidized glutathione, cysteinyl leukotrienes, and activated aflatoxin B1 as substrates. This protein also transports glucuronides and sulfate conjugates of steroid hormones and bile salts. Alternative splicing by exon deletion results in several splice variants but maintains the original open reading frame in all forms. [provided by RefSeq

Specifications

Antigen ABCC1
Applications Flow Cytometry, Immunocytochemistry, Immunoprecipitation
Classification Monoclonal
Clone QCRL-3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant ABCC1.
Dilution Flow Cytometry (1:20-1:50) The optimal working dilution should be determined by the end user.
Gene ABCC1
Gene Alias ABC29/ABCC/DKFZp686N04233/DKFZp781G125/GS-X/MRP/MRP1
Gene Symbols ABCC1
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 617-932 of ABCC1.
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4363
Test Specificity QCRL-3 reacts with a conformation-dependent internal epitope of MRP1. It does not cross-react with human MDR1 and MDR3 gene products nor with murine MRP1. Cross reacts with monkey, but does not cross react with mouse or rat.
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.